Läs mer
TH (phospho S19), Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Beskrivning
The protein encoded by this gene is involved in the conversion of tyrosine to dopamine. It is the rate-limiting enzyme in the synthesis of catecholamines, hence plays a key role in the physiology of adrenergic neurons. Mutations in this gene have been associated with autosomal recessive Segawa syndrome. Alternatively spliced transcript variants encoding different isoforms have been noted for this gene. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | TH |
| Användningsområden | Immunofluorescence |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic phosphopeptide of TH. |
| Utspädning | Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) |
| Gen | TH |
| Gene Alias | TYH |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.