Läs mer
TMC8 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Beskrivning
Epidermodysplasia verruciformis (EV) is an autosomal recessive dermatosis characterized by abnormal susceptibility to human papillomaviruses (HPVs) and a high rate of progression to squamous cell carcinoma on sun-exposed skin. EV is caused by mutations in either of two adjacent genes located on chromosome 17q25.3. Both of these genes encode integral membrane proteins that localize to the endoplasmic reticulum and are predicted to form transmembrane channels. This gene encodes a transmembrane channel-like protein with 8 predicted transmembrane domains and 3 leucine zipper motifs. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | TMC8 |
| Användningsområden | Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of TMC8. |
| Utspädning | Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (0.02% sodium azide) |
| Gen | TMC8 |
| Gene Alias | EV2/EVER2/EVIN2/FLJ40668/FLJ43684/MGC102701/MGC40121 |
| Gensymboler | TMC8 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.