missing translation for 'onlineSavingsMsg'
Läs mer

TRAF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Produktkod. 16088676
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 μL
Förpackningsstorlek:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16088676 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16088676 Leverantör Abnova Leverantörsnummer PAB0274.50uL

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit polyclonal antibody raised against synthetic peptide of TRAF1.

The protein encoded by this gene is a member of the TNF receptor (TNFR) associated factor (TRAF) protein family. TRAF proteins associate with, and mediate the signal transduction from various receptors of the TNFR superfamily. This protein and TRAF2 form a heterodimeric complex, which is required for TNF-alpha-mediated activation of MAPK8/JNK and NF-kappaB. The protein complex formed by this protein and TRAF2 also interacts with inhibitor-of-apoptosis proteins (IAPs), and thus mediates the anti-apoptotic signals from TNF receptors. The expression of this protein can be induced by Epstein-Barr virus (EBV). EBV infection membrane protein 1 (LMP1) is found to interact with this and other TRAF proteins; this interaction is thought to link LMP1-mediated B lymphocyte transformation to the signal transduction from TNFR family receptors. [provided by RefSeq

Sequence: MASSSGSSPRPAPDENEFPFG

Specifikationer

Antigen TRAF1
Användningsområden Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Klassificering Polyclonal
Konjugera Unconjugated
Beskrivning Rabbit polyclonal antibody raised against synthetic peptide of TRAF1.
Utspädning The optimal working dilution should be determined by the end user.
Formulering In serum (0.05% sodium azide)
Gen TRAF1
Gene Alias EBI6/MGC:10353
Gensymboler TRAF1
Värd art Rabbit
Immunogen A synthetic peptide corresponding to amino acids 1-22 of human TRAF1.
Kvantitet 50 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 7185
Målarter Human
Innehåll och lagring Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Visa mer Visa mindre

For Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.