Läs mer
TRAP1, Rabbit, Polyclonal Antibody, Abnova™
Beskrivning
HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. TRAP1 is a mitochondrial HSP90 protein. Other HSP90 proteins are found in cytosol (see HSP90AA1; MIM 140571) and endoplasmic reticulum (HSP90B1; MIM 191175) (Chen et al., 2005 [PubMed 16269234]).[supplied by OMIM
Specifikationer
Specifikationer
| Antigen | TRAP1 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Rabbit polyclonal antibody raised against synthetic peptide of TRAP1. |
| Utspädning | ELISA (1:160000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. |
| Formulering | In buffer containing 0.02% sodium azide |
| Gen | TRAP1 |
| Gene Alias | HSP75/HSP90L |
| Gensymboler | TRAP1 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.