missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trpm2 Rabbit anti-Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Trpm2.
Brand: Abnova PAB11991.100uL
Additional Details : Weight : 0.02000kg
Specifications
Trpm2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of Trpm2. | |
In PBS (30% glycerol, 0.09% sodium azide) | |
9830168K16Rik/C79133/LTRPC2/TRPC7/Trp7/Trrp7 | |
Rabbit | |
100 μL | |
Primary | |
Mouse, Rat | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
Trpm2 | |
Trpm2 | |
A synthetic peptide corresponding to amino acids 1200-1300 of mouse Trpm2. | |
RUO | |
28240 | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only