missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VLDL, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native VLDL.
Brand: Abnova PAB5136.150ug
Additional Details : Weight : 0.02000kg
Specifications
VLDL | |
Polyclonal | |
Rabbit polyclonal antibody raised against native VLDL. | |
In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
Native purified human very low density lipoproteins. | |
RUO | |
Human | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Rabbit | |
150 μg | |
Primary | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only