Läs mer
Rabbit IgG Fab Goat anti-Rabbit, Clone: RMG01 (Biotin), Abnova™
Beskrivning
Goat monoclonal antibody raised against native rabbit IgG Fab.
- High specificity
- High reactivity
Specifikationer
Specifikationer
| Antigen | Rabbit IgG Fab |
| Användningsområden | ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Klassificering | Monoclonal |
| Klona | RMG01 |
| Konjugera | Biotin |
| Beskrivning | Goat monoclonal antibody raised against native rabbit IgG Fab. |
| Utspädning | ELISA (0.01-0.5 ug/mL) Flow Cytometry Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (0.5-2 ug/mL) Immunoprecipitation Western Blotting (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulering | In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) |
| Värd art | Goat |
| Immunogen | Native purified rabbit IgG. |
| Visa mer |
For Research Use Only (RUO)