Läs mer
Rabeprazole Sulfide N-Oxide, TRC
Produktkod. 17940455
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17940455 25 mg
17954295 50 mg
17934375 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17940455 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR07053525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 924663-40-1
Molekylformel C18 H21 N3 O3 S
Molekylvikt (g/mol) 359.44
Synonym 2-[[[4-(3-Methoxypropoxy)-3-methyl-1-oxido-2-pyridinyl]methyl]thio]-1H-benzimidazole, Rabeprazole Sulfide N-Oxide
IUPAC-namn 2-(1H-benzimidazol-2-ylsulfanylmethyl)-4-(3-methoxypropoxy)-3-methylpyridine 1-oxide
LEDER COCCCOc1ccn(=O)c(CSc2nc3ccccc3[nH]2)c1C
Specifikationer
Kemiskt namn eller material Rabeprazole Sulfide N-Oxide
Kvantitet 25 mg
InChI formel InChI=1S/C18H21N3O3S/c1-13-16(21(22)9-8-17(13)24-11-5-10-23-2)12-25-18-19-14-6-3-4-7-15(14)20-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,19,20)
Formel vikt 359.1304