Läs mer
Rabeprazole Sulfide, TRC
Produktkod. 30006756
Change view
Kvantitet:
10 mg
25 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006756 25 mg
30012860 1 g
30007117 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR07051025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 117977-21-6
Molekylformel C18 H21 N3 O2 S
Molekylvikt (g/mol) 343.4432
Synonym 2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]thio]-1H-benzimidazole
IUPAC-namn 2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfanyl]-1H-benzimidazole
LEDER COCCCOc1ccnc(CSc2nc3ccccc3[nH]2)c1C
Specifikationer
Kemiskt namn eller material 2-(((4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl)methyl)thio)-1H-benzimidazole
Kvantitet 25 mg
InChI formel InChI=1S/C18H21N3O2S/c1-13-16(19-9-8-17(13)23-11-5-10-22-2)12-24-18-20-14-6-3-4-7-15(14)21-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,20,21)
Formel vikt 343.1354