missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABGGTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14448-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Catalyzes the transfer of a geranylgeranyl moiety from geranylgeranyl diphosphate to both cysteines of Rab proteins with the C-terminal sequence -XXCC, -XCXC and -CCXX, such as RAB1A, RAB3A, RAB5A and RAB7A.Specifications
RABGGTA | |
Polyclonal | |
Unconjugated | |
RABGGTA | |
PTAR3, Rab GG transferase alpha, Rab GGTase alpha, RABGGTA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
56187, 5875, 58983 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08602, Q92696, Q9JHK4 | |
Rabggta | |
RABGGTA Fusion Protein Ag5923 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |