Läs mer
rac 1,2-Bis-palmitoyl-3-chloropropanediol, TRC
Produktkod. 17948704
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17948704 25 mg
17928604 10 mg
17908434 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17948704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB51520025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 51930-97-3
Molekylformel C35 H67 Cl O4
Molekylvikt (g/mol) 587.36
Synonym Hexadecanoic acid, 1,1'-[1-(chloromethyl)-1,2-ethanediyl] ester, Hexadecanoic acid, 1-(chloromethyl)-1,2-ethanediyl ester (9CI), 3-Chloropropane-1,2-diol dipalmitate, Rac-1,2-bis(palmitoyl)-3-chloropropane-1,2-diol, rac 1,2-Bis-palmitoyl-3-chloropropanediol
IUPAC-namn (3-chloro-2-hexadecanoyloxypropyl) hexadecanoate
LEDER CCCCCCCCCCCCCCCC(=O)OCC(CCl)OC(=O)CCCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material rac-1,2-Bis-palmitol-3-chloropropanediol
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C35H67ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32H2,1-2H3
Formel vikt 586.4728
Procent renhet >95%