missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac 1,2-Dioleoyl-3-chloropropanediol, TRC
Specifications
Specifications
| Kemiskt namn eller material | 1,2-Dioleoyl-3-chloropropanediol |
| CAS | 69161-73-5 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C39 H71 Cl O4 |
| Kvantitet | 25 mg |
| Synonym | 9-Octadecenoic acid (9Z)-, 1-(chloromethyl)-1,2-ethanediyl ester (9CI), 9-Octadecenoic acid (Z)-, 1-(chloromethyl)-1,2-ethanediyl ester, Oleic acid, (chloromethyl)ethylene ester (7CI), 3-Chloro-1,2-propanediol dioleate, MCPD dioleate, Rac-1,2-bis(oleoyl)-3-chloropropane-1,2-diol |
| InChI formel | InChI=1S/C39H71ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37H,3-16,21-36H2,1-2H3/b19-17-,20-18- |
| LEDER | CCCCCCCC\C=C/CCCCCCCC(=O)OCC(CCl)OC(=O)CCCCCCC\C=C/CCCCCCCC |
| IUPAC-namn | [3-chloro-2-[(Z)-octadec-9-enoyl]oxypropyl] (Z)-octadec-9-enoate |
| Molekylvikt (g/mol) | 639.43 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?