Läs mer
rac-1,2-Distearoyl-3-chloropropanediol, TRC
Produktkod. 30133532
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133532 50 mg
30005666 10 mg
30007138 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133532 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD49350050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72468-92-9
Molekylformel C39 H75 Cl O6
Molekylvikt (g/mol) 675.46
Synonym 1,2-Bis-stearoyl-3-chlorpropanediol, Octadecanoic acid, 1-(chloromethyl)-1,2-ethanediyl ester (9CI), Stearic acid, (chloromethyl)ethylene ester (7CI), 3-Chloro-1,2-propanediol distearate, NSC 79288, Rac-1,2-bis(stearoyl)-3-chloropropane-1,2-diol
IUPAC-namn (3-chloro-2-octadecanoylperoxypropyl) octadecaneperoxoate
LEDER CCCCCCCCCCCCCCCCCC(=O)OOCC(CCl)OOC(=O)CCCCCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material rac-1,2-Distearoyl-3-chloropropanediol
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C39H75ClO6/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)45-43-36-37(35-40)44-46-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37H,3-36H2,1-2H3
Formel vikt 674.5252
Procent renhet >95%