Läs mer
rac 1-Methoxy Ketorolac, TRC
Produktkod. 30135939
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30135939 2.5 mg 2.5mg
30135067 25 mg -
30004596 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2627252.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-45-4
Molekylformel C16 H15 N O4
Molekylvikt (g/mol) 285.29
Synonym 5-Benzoyl-2,3-dihydro-1-methoxy-1H-pyrrolizine-1-carboxylic Acid, (1RS)-5-Benzoyl-1-methoxy-2,3-dihydro-1H-pyrrolizine-1-carboxylic Acid
IUPAC-namn 5-benzoyl-1-methoxy-2,3-dihydropyrrolizine-1-carboxylic acid
LEDER COC1(CCn2c(ccc12)C(=O)c3ccccc3)C(=O)O
Specifikationer
Kemiskt namn eller material rac 1-Methoxy Ketorolac
Kvantitet 2.5 mg
InChI formel InChI=1S/C16H15NO4/c1-21-16(15(19)20)9-10-17-12(7-8-13(16)17)14(18)11-5-3-2-4-6-11/h2-8H,9-10H2,1H3,(H,19,20)
Formel vikt 285.1001