Läs mer
rac-1-Palmitoyl-2-linolenoyl-3-chloropropanediol, TRC
Produktkod. 30138229
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138229 25 mg
30009070 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP15885025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1246833-14-6
Molekylformel C37H65ClO4
Molekylvikt (g/mol) 609.36
Synonym 9,12,15-Octadecatrienoic Acid 1-(Chloromethyl)-2-[(1-oxohexadecyl)oxy]ethyl Ester, Palmitic-linolenic MCPD;
IUPAC-namn [(2S)-1-chloro-3-hexadecanoyloxypropan-2-yl] (9Z,12Z,15E)-octadeca-9,12,15-trienoate
LEDER O=C(CCCCCCCCCCCCCCC)OC[C@H](OC(CCCCCCC/C=C\C/C=C\C/C=C/CC)=O)CCl
Specifikationer
Kemiskt namn eller material rac-1-Palmitoyl-2-linolenoyl-3-chloropropanediol
Kvantitet 25 mg
InChI formel InChI=1S/C37H65ClO4/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-37(40)42-35(33-38)34-41-36(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h5,7,11,13,17-18,35H,3-4,6,8-10,12,14-16,19-34H2,1-2H3/b7-5+,13-11-,18-17-/t35-/m1/s1
Formel vikt 608.4571