Läs mer
rac 1-Palmitoyl-2-stearoyl-3-chloropropanediol, TRC
Produktkod. 30411977
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411977 25 mg
30424851 10 mg
30414494 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411977 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP16350025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1185060-41-6
Molekylformel C37 H71 Cl O4
Molekylvikt (g/mol) 615.41
IUPAC-namn 18-(chloromethyl)-17,18-dihydroxyhexatriacontane-16,19-dione
LEDER CCCCCCCCCCCCCCCCCC(=O)C(O)(CCl)C(O)C(=O)CCCCCCCCCCCCCCC
Specifikationer
Kemiskt namn eller material 1-Palmitoyl-2-stearoyl-3-chloropropanediol
Kvantitet 25 mg
InChI formel InChI=1S/C37H71ClO4/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-35(40)37(42,33-38)36(41)34(39)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2/h36,41-42H,3-33H2,1-2H3
Formel vikt 614.5041