Läs mer
rac 2-Bromo Phenylephrine Hydrochloride, TRC
Produktkod. 30339347
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30339347 10 mg
1 options
Produktkod. 30339347 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68681510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (R)-2-Bromo Phenylephrine Hydrochloride
CAS 1391054-09-3
Molekylformel C9H13BrClNO2
Kvantitet 10 mg
Synonym Rac 2-Bromo Phenylephrine Hydrochloride
InChI formel InChI=1S/C9H12BrNO2.ClH/c1-11-5-8(13)6-3-2-4-7(12)9(6)10;/h2-4,8,11-13H,5H2,1H3;1H/t8-;/m0./s1
LEDER Cl.CNC[C@H](O)c1cccc(O)c1Br
IUPAC-namn 2-bromo-3-[(1R)-1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
Molekylvikt (g/mol) 282.56
Formel vikt 280.9818