Läs mer
rac 2-Palmitoyl-3-chloropropanediol, TRC
Produktkod. 30414928
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30414928 50 mg
30423210 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30414928 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP15651050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac 2-Palmitoyl-3-chloropropanediol
CAS 20618-92-2
Molekylformel C19H37ClO3
Kvantitet 50 mg
Synonym Hexadecanoic Acid 2-Chloro-1-(hydroxymethyl)ethyl Ester, Palmitic Acid 2-Chloro-1-(hydroxymethyl)ethyl Ester, 3-Chloro-1,2-propanediol 2-Palmitate
InChI formel InChI=1S/C19H37ClO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(22)23-18(16-20)17-21/h18,21H,2-17H2,1H3
LEDER O=C(CCCCCCCCCCCCCCC)OC(CCl)CO
IUPAC-namn (1-chloro-3-hydroxypropan-2-yl) hexadecanoate
Molekylvikt (g/mol) 348.95
Formel vikt 348.2431