Läs mer
rac 3,4-Dihydroxyphenylethylene Glycol, TRC
Produktkod. 30133703
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133703 25 mg
30133239 100 mg
30003307 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133703 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD45451025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 28822-73-3
Molekylformel C8 H10 O4
Molekylvikt (g/mol) 170.16
Synonym DL-3,4-Dihydroxyphenylglycol, (+/-)-4-(1,2-Dihydroxyethyl)pyrocatechol, 4-(1,2-Dihydroxyethyl)-1,2-benzenediol
IUPAC-namn 4-(1,2-dihydroxyethyl)benzene-1,2-diol
LEDER OCC(O)c1ccc(O)c(O)c1
Specifikationer
Kemiskt namn eller material rac 3,4-Dihydroxyphenylethylene Glycol
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C8H10O4/c9-4-8(12)5-1-2-6(10)7(11)3-5/h1-3,8-12H,4H2
Formel vikt 170.0579
Procent renhet >95%