Läs mer
rac-3-Octadecanamido-2-ethoxypropan-1-ol Phosphocholine, TRC
Produktkod. 30420734
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30420734 25 mg
1 options
Produktkod. 30420734 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO23400025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-3-Octadecanamido-2-Ethoxypropan-1-ol Phosphocholine
CAS 112989-02-3
Molekylformel C28 H59 N2 O6 P
Kvantitet 25 mg
Synonym 3,5-Dioxa-9-aza-4-phosphaheptacosan-1-aminium','7-ethoxy-4-hydroxy-N,N,N-trimethyl-10-oxo-','inner salt','4-oxide, 3,5-Dioxa-9-aza-4-phosphaheptacosan-1-aminium','7-ethoxy-4-hydroxy-N,N,N-trimethyl-10-oxo-','inner salt','4-oxide','(±)-, CP 51, rac-3-Octadecanamido-2-ethoxypropan-1-ol Phosphocholine
InChI formel InChI=1S/C28H59N2O6P/c1-6-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-28(31)29-25-27(34-7-2)26-36-37(32,33)35-24-23-30(3,4)5/h27H,6-26H2,1-5H3,(H-,29,31,32,33)
LEDER CCCCCCCCCCCCCCCCCC(=O)NCC(COP(=O)([O-])OCC[N+](C)(C)C)OCC
IUPAC-namn [2-ethoxy-3-(octadecanoylamino)propyl] 2-(trimethylazaniumyl)ethyl phosphate
Molekylvikt (g/mol) 550.75
Formel vikt 550.4111