Läs mer
rac 4,5-Dehydro Pregabalin, TRC
Produktkod. 30011092
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011092 50 mg
30003790 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011092 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23005050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 216576-74-8
Molekylformel C8 H15 N O2
Molekylvikt (g/mol) 157.21
Synonym 4-Hexenoic acid, 3-(aminomethyl)-5-methyl-, 3-(Aminomethyl)-5-methyl-4-hexenoic acid
IUPAC-namn 3-(aminomethyl)-5-methylhex-4-enoic acid
LEDER CC(=CC(CN)CC(=O)O)C
Specifikationer
Kemiskt namn eller material rac 4,5-Dehydro Pregabalin
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C8H15NO2/c1-6(2)3-7(5-9)4-8(10)11/h3,7H,4-5,9H2,1-2H3,(H,10,11)
Kvantitet 50 mg
Formel vikt 157.1103
Procent renhet >95%