Läs mer
rac 4-Bromo Phenylephrine Hydrochloride, TRC
Produktkod. 30332265
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30332265 50 mg
1 options
Produktkod. 30332265 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB68682050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac 4-Bromo Phenylephrine Hydrochloride
CAS 1391054-36-6
Molekylformel C9H13BrClNO2
Kvantitet 50 mg
Synonym 4-Bromo-3-hydroxy-alpha-[(methylamino)methyl]benzenemethanol Hydrochloride, 4-Bromophenylephrine Hydrochloride
InChI formel InChI=1S/C9H12BrNO2.ClH/c1-11-5-9(13)6-2-3-7(10)8(12)4-6;/h2-4,9,11-13H,5H2,1H3;1H
LEDER Cl.CNCC(O)c1ccc(Br)c(O)c1
IUPAC-namn 2-bromo-5-[1-hydroxy-2-(methylamino)ethyl]phenol;hydrochloride
Molekylvikt (g/mol) 282.56
Formel vikt 280.9818