Läs mer
rac 4-Hydroxy-3-methoxyphenylethylene Glycol, TRC
Produktkod. 30010668
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010668 10 mg
30005998 25 mg
30004539 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010668 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94634010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 534-82-7
Molekylformel C9H12O4
Molekylvikt (g/mol) 184.19
IUPAC-namn 1-(4-hydroxy-3-methoxyphenyl)ethane-1,2-diol
LEDER COc1cc(ccc1O)C(O)CO
Specifikationer
Kemiskt namn eller material (±)-4-Hydroxy-3-methoxyphenylethylene Glycol (MHPG)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C9H12O4/c1-13-9-4-6(8(12)5-10)2-3-7(9)11/h2-4,8,10-12H,5H2,1H3
Formel vikt 184.0736
Procent renhet >95%