Läs mer
rac-4-Hydroxy Propranolol Hydrochloride, TRC
Produktkod. 30009485
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009485 5 mg
30007078 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9525455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14133-90-5
Molekylformel C16 H21 N O3 . Cl H
Molekylvikt (g/mol) 311.8038
Synonym 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (9CI), 2-Propanol, 1-[(4-hydroxy-1-naphthyl)oxy]-3-(isopropylamino)-, hydrochloride (8CI), 4-Hydroxypropranolol hydrochloride, 1-Naphthalenol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-, hydrochloride (1:1), 4-Hydroxypropranolol Hydrochloride
IUPAC-namn 4-[2-hydroxy-3-(propan-2-ylamino)propoxy]naphthalen-1-ol;hydrochloride
LEDER Cl.CC(C)NCC(O)COc1ccc(O)c2ccccc12
Specifikationer
Kemiskt namn eller material (+/-)-4-Hydroxy Propranolol Hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C16H21NO3.ClH/c1-11(2)17-9-12(18)10-20-16-8-7-15(19)13-5-3-4-6-14(13)16;/h3-8,11-12,17-19H,9-10H2,1-2H3;1H
Formel vikt 311.1288
Procent renhet >95%