Läs mer
rac 5,6-Dehydro Pregabalin, TRC
Produktkod. 17930355
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17930355 1 mg
17922495 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17930355 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2300551MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1136478-30-2
Molekylformel C8H15NO2
Molekylvikt (g/mol) 157.21
Synonym 5-Hexenoic acid, 3-(aminomethyl)-5-methyl-, 3-(Aminomethyl)-5-methyl-5-hexenoic acid, 3-(Aminomethyl)-5-methylhex-5-enoic acid, (RS)-5,6-Dehydropregabalin, rac 5,6-Dehydro Pregabalin
LEDER CC(=C)CC(CN)CC(=O)O
Specifikationer
Kemiskt namn eller material rac 5,6-Dehydro Pregabalin
InChI formel InChI=1S/C8H15NO2/c1-6(2)3-7(5-9)4-8(10)11/h7H,1,3-5,9H2,2H3,(H,10,11)
Kvantitet 1 mg
Formel vikt 157.1103