Läs mer
rac 5-Carboxy Desisopropyl Tolterodine Methyl Ester, TRC
Produktkod. 30358617
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30358617 5 mg
1 options
Produktkod. 30358617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1779355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac 5-Carboxy Desisopropyl Tolterodine Methyl Ester
CAS 1391053-28-3
Molekylformel C20H25NO3
Kvantitet 5 mg
Synonym 4-Hydroxy-3-[3-[(1-methylethyl)amino]-1-phenylpropyl]-benzoic Acid Methyl Ester
InChI formel InChI=1S/C20H25NO3/c1-14(2)21-12-11-17(15-7-5-4-6-8-15)18-13-16(20(23)24-3)9-10-19(18)22/h4-10,13-14,17,21-22H,11-12H2,1-3H3
LEDER COC(=O)c1ccc(O)c(c1)C(CCNC(C)C)c2ccccc2
IUPAC-namn methyl 4-hydroxy-3-[1-phenyl-3-(propan-2-ylamino)propyl]benzoate
Molekylvikt (g/mol) 327.42
Formel vikt 327.1834