Läs mer
rac 5-Desfluoro Voriconazole, TRC
Produktkod. 30364136
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30364136 1 mg
30361699 5 mg
30360898 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30364136 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2898701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 182369-73-9
Molekylformel C16 H15 F2 N5 O
Molekylvikt (g/mol) 331.32
Synonym 4-Pyrimidineethanol, α-(2,4-difluorophenyl)-β-methyl-α-(1H-1,2,4-triazol-1-ylmethyl)-, (R*,S*)- (9CI), (2RS,3SR)-2-(2,4-Di-fluorophenyl)-3-pyrimidin-4-yl-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, Voriconazole Imp. B (EP)
IUPAC-namn (2R,3S)-2-(2,4-difluorophenyl)-3-pyrimidin-4-yl-1-(1,2,4-triazol-1-yl)butan-2-ol
LEDER C[C@@H](c1ccncn1)[C@](O)(Cn2cncn2)c3ccc(F)cc3F
Specifikationer
Kemiskt namn eller material (2RS,3SR)-2-(2,4-Difluorophenyl)-3-pyrimidin-4-yl-1-(1H-1,2,4-triazol-1-yl)butan-2-ol
Kvantitet 1 mg
InChI formel InChI=1S/C16H15F2N5O/c1-11(15-4-5-19-8-21-15)16(24,7-23-10-20-9-22-23)13-3-2-12(17)6-14(13)18/h2-6,8-11,24H,7H2,1H3/t11-,16+/m0/s1
Formel vikt 331.1245