Läs mer
rac-6-Methyl-4-phenyl-2-chromanol (Tolterodine Impurity) (Mixture of Diastereomers), TRC
Produktkod. 30135629
Change view
Kvantitet:
100 mg
250 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135629 2.5 g
30136051 100 mg
30009968 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135629 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3270702.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 209747-04-6
Molekylformel C16 H16 O2
Molekylvikt (g/mol) 240.3
Synonym 2H-1-Benzopyran-2-ol, 3,4-dihydro-6-methyl-4-phenyl-, 3,4-Dihydro-6-methyl-4-phenyl-2H-1-benzopyran-2-ol, 3,4-Dihydro-6-methyl-4-phenyl-2H-benzopyran-2-ol, 6-Methyl-4-phenyl-2-chromanol, (4RS)-6-Methyl-4-phenylchroman-2-ol, Tolterodine Impurity
IUPAC-namn 6-methyl-4-phenyl-3,4-dihydro-2H-chromen-2-ol
LEDER Cc1ccc2OC(O)CC(c3ccccc3)c2c1
Specifikationer
Kemiskt namn eller material rac-6-Methyl-4-phenyl-2-chromanol (Mixture of Diastereomers)
Kvantitet 2.5 g
InChI formel InChI=1S/C16H16O2/c1-11-7-8-15-14(9-11)13(10-16(17)18-15)12-5-3-2-4-6-12/h2-9,13,16-17H,10H2,1H3
Formel vikt 240.115