Läs mer
Rac 8’-Acetylene Abscisic Acid Methyl Ester, TRC
Produktkod. 30313639
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30313639 25 mg
1 options
Produktkod. 30313639 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA11001025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac 8'-Acetylene Abscisic Acid Methyl Ester
CAS 192987-96-5
Molekylformel C17 H20 O4
Kvantitet 25 mg
Synonym (2Z,4E)-rel-5-[(1R,6R)-6-ethynyl-1-hydroxy-2,6-dimethyl-4-oxo-2-cyclohexen-1-yl]-3-methyl-2,4-pentadienoic Acid Methyl Ester, PBI 429
InChI formel InChI=1S/C17H20O4/c1-6-16(4)11-14(18)10-13(3)17(16,20)8-7-12(2)9-15(19)21-5/h1,7-10,20H,11H2,2-5H3/b8-7+,12-9-/t16-,17+/m0/s1
LEDER COC(=O)\C=C(\C)/C=C/[C@@]1(O)C(=CC(=O)C[C@]1(C)C#C)C
IUPAC-namn methyl (2Z,4E)-5-[(1R,6R)-6-ethynyl-1-hydroxy-2,6-dimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoate
Molekylvikt (g/mol) 288.338
Formel vikt 288.136