Läs mer
rac 8-Hydroxy Efavirenz, TRC
Produktkod. 30137953
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137953 10 mg
30136150 1 mg
30009982 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94182510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 205754-32-1
Molekylformel C14 H9 Cl F3 N O3
Molekylvikt (g/mol) 331.67
Synonym 2H-3,1-Benzoxazin-2-one, 6-chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-8-hydroxy-4-(trifluoromethyl)-, 2H-3,1-Benzoxazin-2-one, 6-chloro-4-(cyclopropylethynyl)-1,4-dihydro-8-hydroxy-4-(trifluoromethyl)- (9CI), 6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-8-hydroxy-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, rac 8-Hydroxy Efavirenz
IUPAC-namn 6-chloro-4-(2-cyclopropylethynyl)-8-hydroxy-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one
LEDER Oc1cc(Cl)cc2c1NC(=O)OC2(C#CC3CC3)C(F)(F)F
Specifikationer
Kemiskt namn eller material rac 8-Hydroxy efavirenz
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C14H9ClF3NO3/c15-8-5-9-11(10(20)6-8)19-12(21)22-13(9,14(16,17)18)4-3-7-1-2-7/h5-7,20H,1-2H2,(H,19,21)
Formel vikt 331.0223
Procent renhet >95%