Läs mer
rac Atomoxetine Hydrochloride, TRC
Produktkod. 30333300
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30333300 5 mg
1 options
Produktkod. 30333300 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7913905MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-Atomoxetine hydrochloride
CAS 82857-40-7
Molekylformel C17 H21 N O . Cl H
Kvantitet 5 mg
Synonym Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride (1:1), Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride (9CI), Benzenepropanamine, N-methyl-γ-(2-methylphenoxy)-, hydrochloride, (±)-, LY 135252, N-Methyl-3-(2-methylphenoxy)-3-phenylpropanamine hydrochloride, N-Methyl-3-(2-methylphenoxy)-3-phenylpropylamine hydrochloride, rac Atomoxetine Hydrochloride, (RS)-Atomoxetine Hydrochloride
InChI formel InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H
LEDER Cl.CNCCC(Oc1ccccc1C)c2ccccc2
IUPAC-namn N-methyl-3-(2-methylphenoxy)-3-phenylpropan-1-amine;hydrochloride
Molekylvikt (g/mol) 291.82
Formel vikt 291.139