Läs mer
rac cis-Moxifloxacin Acyl-b-D-glucuronide (>85%), TRC
Produktkod. 30474461
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474461 10 mg
30474318 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30474461 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM74501010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Moxifloxacin Acyl-beta-D-glucuronide
CAS 733002-61-4
Molekylformel C27 H32 F N3 O10
Kvantitet 10 mg
Synonym Rac Cis-Moxifloxacin Acyl-B-D-Glucuronide
InChI formel InChI=1S/C27H32FN3O10/c1-39-23-17-13(7-15(28)18(23)30-8-11-3-2-6-29-16(11)10-30)19(32)14(9-31(17)12-4-5-12)26(38)41-27-22(35)20(33)21(34)24(40-27)25(36)37/h7,9,11-12,16,20-22,24,27,29,33-35H,2-6,8,10H2,1H3,(H,36,37)/t11-,16+,20-,21-,22+,24-,27-/m0/s1
LEDER COc1c(N2C[C@@H]3CCCN[C@@H]3C2)c(F)cc4C(=O)C(=CN(C5CC5)c14)C(=O)O[C@@H]6O[C@@H]([C@@H](O)[C@H](O)[C@H]6O)C(=O)O
IUPAC-namn (2S,3S,4S,5R,6S)-6-[7-[(4aS,7aS)-1,2,3,4,4a,5,7,7a-octahydropyrrolo[3,4-b]pyridin-6-yl]-1-cyclopropyl-6-fluoro-8-methoxy-4-oxoquinoline-3-carbonyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
Molekylvikt (g/mol) 577.56
Formel vikt 577.2072