Läs mer
rac-Clopidogrel Carboxylic Acid, TRC
Produktkod. 30137989
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137989 500 mg
30133963 1 g
17905005 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137989 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC587241500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 90055-55-3
Molekylformel C15 H14 Cl N O2 S
Molekylvikt (g/mol) 307.8
Synonym (R,S)-Clopidogrel (free acid), rac-Clopidogrel carboxylic acid, Thieno[3,2-c]pyridine-5(4H)-acetic acid, α-(2-chlorophenyl)-6,7-dihydro-, α-(2-Chlorophenyl)-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-acetic acid, (±)-(2-Chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetic acid, 2-(2-Chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid, 2-(2-Chlorophenyl)-2-[6,7-dihydrothieno[3,2-c]pyridine-5(4H)-yl]acetic acid, (2RS)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic acid, (2RS)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]ethanoic acid, (RS)-Clopidogrel acid
IUPAC-namn 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid
LEDER OC(=O)C(N1CCc2sccc2C1)c3ccccc3Cl
Specifikationer
Kemiskt namn eller material (RS)-Clopidogrel Carboxylic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C15H14ClNO2S/c16-12-4-2-1-3-11(12)14(15(18)19)17-7-5-13-10(9-17)6-8-20-13/h1-4,6,8,14H,5,7,9H2,(H,18,19)
Formel vikt 307.0434
Procent renhet >95%