Läs mer
rac-Clopidogrel Hydrogen Sulfate, TRC
Produktkod. 30136148
Change view
Kvantitet:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136148 500 mg
30009133 25 mg
17954615 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136148 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC587248500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 135046-48-9
Molekylformel C16 H16 Cl N O2 S . H2 O4 S
Molekylvikt (g/mol) 419.9
Synonym rac-Clopidogrel bisulfate, (RS)-Methyl (2-chlorophenyl)(6,7-dihydro-4H-thieno[3,2-c]pyrid-5-yl)acetate hydrogen sulfate salt, Methyl 2-(2-Chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate hydrogen sulfate, (RS)-Clopidogrel Hydrogen Sulphate
IUPAC-namn methyl 2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
LEDER COC(=O)C(N1CCc2sccc2C1)c3ccccc3Cl.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material rac-Clopidogrel Hydrogen Sulfate
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C16H16ClNO2S.H2O4S/c1-20-16(19)15(12-4-2-3-5-13(12)17)18-8-6-14-11(10-18)7-9-21-14;1-5(2,3)4/h2-5,7,9,15H,6,8,10H2,1H3;(H2,1,2,3,4)
Formel vikt 419.0264
Procent renhet >95%