Läs mer
rac-Clopidogrel-MP Endo Derivative, TRC
Produktkod. 30353488
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30353488 25 mg
1 options
Produktkod. 30353488 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC58728025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(1-(1-(2-chlorophenyl)-2-methoxy-2-oxoethyl)-4-((2-(3-methoxyphenyl)-2-oxoethyl)thio)-1,2,5,6-tetrahydropyridin-3-yl)acetic acid
CAS 1346598-12-6
Molekylformel C25H26ClNO6S
Kvantitet 25 mg
Synonym 3,4-Dehydro-3-(carboxymethyl)-α-(2-chlorophenyl)-4-[[2-(3-methoxyphenyl)-2-oxoethyl]thio]-1-piperidineacetic Acid 1-Methyl Ester, rac-Clopidogrel-MP Endo Derivative
InChI formel InChI=1S/C25H26ClNO6S/c1-32-18-7-5-6-16(12-18)21(28)15-34-22-10-11-27(14-17(22)13-23(29)30)24(25(31)33-2)19-8-3-4-9-20(19)26/h3-9,12,24H,10-11,13-15H2,1-2H3,(H,29,30)
LEDER ClC1=CC=CC=C1C(C(OC)=O)N2CCC(SCC(C3=CC(OC)=CC=C3)=O)=C(CC(O)=O)C2
IUPAC-namn 2-(1-(1-(2-chlorophenyl)-2-methoxy-2-oxoethyl)-4-((2-(3-methoxyphenyl)-2-oxoethyl)thio)-1,2,5,6-tetrahydropyridin-3-yl)acetic acid
Molekylvikt (g/mol) 504
Formel vikt 503.12