Läs mer
rac-Des(4-cyclohexyl-L-proline) Fosinopril Acetic Acid(Mixture of Diastereomers) (>80%), TRC
Produktkod. 30351619
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30351619 100 mg
1 options
Produktkod. 30351619 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD288605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-Des(4-cyclohexyl-L-proline) Fosinopril Acetic Acid (Mixture of Diastereomers)
CAS 123599-78-0
Molekylformel C19H29O6P
Kvantitet 100 mg
Synonym rac-2-[[2-Methyl-1-(1-oxopropoxy)propoxy](4-phenylbutyl)phosphinyl]acetic Acid
InChI formel InChI=1S/C19H29O6P/c1-4-18(22)24-19(15(2)3)25-26(23,14-17(20)21)13-9-8-12-16-10-6-5-7-11-16/h5-7,10-11,15,19H,4,8-9,12-14H2,1-3H3,(H,20,21)
LEDER CCC(=O)OC(OP(=O)(CCCCc1ccccc1)CC(=O)O)C(C)C
IUPAC-namn 2-[(2-methyl-1-propanoyloxypropoxy)-(4-phenylbutyl)phosphoryl]acetic acid
Molekylvikt (g/mol) 384.4
Formel vikt 384.1702