Läs mer
rac Desethyl Oxybutynin Hydrochloride, TRC
Produktkod. 30363492
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363492 50 mg
30352206 25 mg
30356009 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363492 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28947550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 81039-77-2
Molekylformel C20 H27 N O3 . Cl H
Molekylvikt (g/mol) 365.89
Synonym Desethyloxybutynin hydrochloride, Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylamino)-2-butyn-1-yl ester, hydrochloride (1:1) (ACI), Benzeneacetic acid, α-cyclohexyl-α-hydroxy-, 4-(ethylamino)-2-butynyl ester, hydrochloride (9CI), α-Cyclohexyl-α-hydroxybenzeneacetic acid 4-(ethylamino)-2-butyn-1-yl ester hydrochloride, 4-(Ethylamino)but-2-yn-1-yl 2-cyclohexyl-2-hydroxy-2-phenylacetate hydrochloride, N-Desethyloxybutynin hydrochloride, N-Desethyl Oxybutynin hydrochloride, rac Desethyl Oxybutynin hydrochloride
IUPAC-namn 4-(ethylamino)but-2-ynyl 2-cyclohexyl-2-hydroxy-2-phenylacetate;hydrochloride
LEDER Cl.CCNCC#CCOC(=O)C(O)(C1CCCCC1)c2ccccc2
Specifikationer
Kemiskt namn eller material Desethyloxybutynin Hydrochloride
Kvantitet 50 mg
InChI formel InChI=1S/C20H27NO3.ClH/c1-2-21-15-9-10-16-24-19(22)20(23,17-11-5-3-6-12-17)18-13-7-4-8-14-18;/h3,5-6,11-12,18,21,23H,2,4,7-8,13-16H2,1H3;1H
Formel vikt 365.1758