Läs mer
rac Diacetolol, TRC
Produktkod. 30137958
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137958 1 g
30007477 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137958 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD3052001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22568-64-5
Molekylformel C16 H24 N2 O4
Molekylvikt (g/mol) 308.37
Synonym Acetamide, N-[3-acetyl-4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]-, Acetamide, N-[3-acetyl-4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]-, (±)-, Acetanilide, 3'-acetyl-4'-[2-hydroxy-3-(isopropylamino)propoxy]-, DL- (8CI), N-[3-Acetyl-4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetamide, (±)-Diacetolol, 1-(4-Acetamido-2-acetylphenoxy)-2-hydroxy-3-isopropylaminopropane, Acetylacebutolol, DL-1-(2-Acetyl-4-acetamidophenoxy)-2-hydroxy-3-isopropylaminopropane, Diacetol, Diacetolol, M and B 16942
IUPAC-namn N-[3-acetyl-4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetamide
LEDER CC(C)NCC(O)COc1ccc(NC(=O)C)cc1C(=O)C
Specifikationer
Kemiskt namn eller material rac Diacetolol
Kvantitet 1 g
InChI formel InChI=1S/C16H24N2O4/c1-10(2)17-8-14(21)9-22-16-6-5-13(18-12(4)20)7-15(16)11(3)19/h5-7,10,14,17,21H,8-9H2,1-4H3,(H,18,20)
Formel vikt 308.1736