Läs mer
rac Dropropizine, TRC
Produktkod. 30134632
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134632 10 g
17901355 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134632 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD68149510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17692-31-8
Molekylformel C13 H20 N2 O2
Molekylvikt (g/mol) 236.31
Synonym Dropropizine
IUPAC-namn 3-(4-phenylpiperazin-1-yl)propane-1,2-diol
LEDER OCC(O)CN1CCN(CC1)c2ccccc2
Specifikationer
Kemiskt namn eller material Dropropizine
Kvantitet 10 g
InChI formel InChI=1S/C13H20N2O2/c16-11-13(17)10-14-6-8-15(9-7-14)12-4-2-1-3-5-12/h1-5,13,16-17H,6-11H2
Formel vikt 236.1525