Läs mer
rac- Etomoxir, TRC
Produktkod. 30368839
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30368839 25 mg
30374509 10 mg
30374700 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30368839 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE93350025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 82258-36-4
Molekylformel C17 H23 Cl O4
Molekylvikt (g/mol) 326.82
IUPAC-namn ethyl 2-[6-(4-chlorophenoxy)hexyl]oxirane-2-carboxylate
LEDER CCOC(=O)C1(CCCCCCOc2ccc(Cl)cc2)CO1
Specifikationer
Kemiskt namn eller material Etomoxir
Kvantitet 25 mg
InChI formel InChI=1S/C17H23ClO4/c1-2-20-16(19)17(13-22-17)11-5-3-4-6-12-21-15-9-7-14(18)8-10-15/h7-10H,2-6,11-13H2,1H3
Formel vikt 326.1285