Läs mer
rac Guaifenesin Cyclic Carbonate, TRC
Produktkod. 30133824
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133824 1 g
30135151 500 mg
17973975 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133824 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG8105151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2049-21-0
Molekylformel C11 H12 O5
Molekylvikt (g/mol) 224.21
Synonym 1,3-Dioxolan-2-one, 4-[(2-methoxyphenoxy)methyl]-, Carbonic acid, cyclic [(o-methoxyphenoxy)methyl]ethylene ester (6CI,7CI,8CI), 1,2-Propanediol, 3-(o-methoxyphenoxy)-, cyclic carbonate (8CI), 4-[(2-Methoxyphenoxy)methyl]-1,3-dioxolan-2-one
IUPAC-namn 4-[(2-methoxyphenoxy)methyl]-1,3-dioxolan-2-one
LEDER COc1ccccc1OCC2COC(=O)O2
Specifikationer
Kemiskt namn eller material rac Guaifenesin Cyclic Carbonate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C11H12O5/c1-13-9-4-2-3-5-10(9)14-6-8-7-15-11(12)16-8/h2-5,8H,6-7H2,1H3
Formel vikt 224.0685
Procent renhet >95%