Läs mer
rac Ketoroloc Tris Amide Impurity, TRC
Produktkod. 30138059
Change view
Kvantitet:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138059 1 g
30136317 250 mg
17983805 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138059 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK2356401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 167105-80-8
Molekylformel C19 H22 N2 O5
Molekylvikt (g/mol) 358.39
Synonym 5-Benzoyl-2,3-dihydro-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]-1H-pyrrolizine-1-carboxamide, 1H-Pyrrolizine-1-carboxamide, 5-benzoyl-2,3-dihydro-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]-, (±)-, (1RS)-5-Benzoyl-N-[2-hydroxy-1,1-bis(hydroxymethyl)ethyl]-2,3-dihydro-1H-pyrrolizine-1-carboxamide
IUPAC-namn 5-benzoyl-N-[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]-2,3-dihydro-1H-pyrrolizine-1-carboxamide
LEDER OCC(CO)(CO)NC(=O)C1CCn2c1ccc2C(=O)c3ccccc3
Specifikationer
Kemiskt namn eller material 5-benzoyl-N-(1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)-2,3-dihydro-1H-pyrrolizine-1-carboxamide
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C19H22N2O5/c22-10-19(11-23,12-24)20-18(26)14-8-9-21-15(14)6-7-16(21)17(25)13-4-2-1-3-5-13/h1-7,14,22-24H,8-12H2,(H,20,26)
Formel vikt 358.1529
Procent renhet >95%