Läs mer
rac Kynurenine, TRC
Produktkod. 30136712
Change view
Kvantitet:
5 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136712 25 mg
30005633 5 mg
30011560 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136712 Leverantör Toronto Research Chemicals Leverantörsnummer TRCK66100025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 343-65-7
Molekylformel C10 H12 N2 O3
Molekylvikt (g/mol) 208.21
Synonym α,2-Diamino-γ-oxobenzenebutanoic Acid, 3-Anthraniloylalanine, DL-Kynurenine, Kynurenin, dl-Kynurenine;
IUPAC-namn 2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
LEDER NC(CC(=O)c1ccccc1N)C(=O)O
Specifikationer
Kemiskt namn eller material rac Kynurenine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8H,5,11-12H2,(H,14,15)
Formel vikt 208.0848
Procent renhet >95%