Läs mer
rac-Lasofoxifene, TRC
Produktkod. 30472089
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30472089 1 mg
1 options
Produktkod. 30472089 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL1764901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac-Lasofoxifene
CAS 180915-78-0
Molekylformel C28H31NO2
Kvantitet 1 mg
Synonym (5R,6S)-rel-5,6,7,8-Tetrahydro-6-phenyl-5-[4-[2-(1-pyrrolidinyl)ethoxy]phenyl]-2-naphthalenol, cis-5,6,7,8-Tetrahydro-6-phenyl-5-[4-[2-(1-pyrrolidinyl)ethoxy]phenyl]-2-naphthalenol
InChI formel InChI=1S/C28H31NO2/c30-24-11-15-27-23(20-24)10-14-26(21-6-2-1-3-7-21)28(27)22-8-12-25(13-9-22)31-19-18-29-16-4-5-17-29/h1-3,6-9,11-13,15,20,26,28,30H,4-5,10,14,16-19H2/t26-,28+/m1/s1
LEDER C1CCN(C1)CCOC2=CC=C(C=C2)[C@H]3[C@H](CCC4=C3C=CC(=C4)O)C5=CC=CC=C5
Molekylvikt (g/mol) 413.55