Läs mer
rac Metanephrine Hydrochloride Salt, TRC
Produktkod. 18033940
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18033940 100 mg
18071750 50 mg
17919649 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18033940 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM258760100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 881-95-8
Molekylformel C10 H15 N O3 . Cl H
Molekylvikt (g/mol) 233.69
Synonym Benzenemethanol, 4-hydroxy-3-methoxy-α-[(methylamino)methyl]-, hydrochloride (1:1), Benzenemethanol, 4-hydroxy-3-methoxy-α-[(methylamino)methyl]-, hydrochloride (9CI), Benzenemethanol, 4-hydroxy-3-methoxy-α-[(methylamino)methyl]-, hydrochloride, (±)-, Vanillyl alcohol, α-[(methylamino)methyl]-, hydrochloride, (±)- (8CI), (±)-Metanephrine hydrochloride, Metanephrine hydrochloride, dl-Metanephrine hydrochloride, D,L-Metanephrine Hydrochloride, rac-Metanephrine Hydrochloride
IUPAC-namn 4-[1-hydroxy-2-(methylamino)ethyl]-2-methoxyphenol;hydrochloride
LEDER Cl.CNCC(O)c1ccc(O)c(OC)c1
Specifikationer
Kemiskt namn eller material D,L-Metanephrine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C10H15NO3.ClH/c1-11-6-9(13)7-3-4-8(12)10(5-7)14-2;/h3-5,9,11-13H,6H2,1-2H3;1H
Formel vikt 233.0819
Procent renhet >95%