Läs mer
rac Methotrimeprazine Maleate Salt, TRC
Produktkod. 30465778
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465778 10 mg
30469113 25 mg
30467563 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30465778 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM26078810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17086-29-2
Molekylformel C23H28N2O5S
Molekylvikt (g/mol) 444.54
Synonym 2-Methoxy-N,N,β-trimethyl-10H-phenothiazine-10-propanamine Maleate, 10-[3-(dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine Maleate, 2-Methoxytrimeprazine Maleate
IUPAC-namn (Z)-but-2-enedioic acid;3-(2-methoxyphenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
LEDER CC(CN1C2=CC=CC=C2SC3=C1C=C(C=C3)OC)CN(C)C.C(=C\C(=O)O)\C(=O)O
Specifikationer
Kemiskt namn eller material rac Methotrimeprazine Maleate Salt
Kvantitet 10 mg
InChI formel InChI=1S/C19H24N2OS.C4H4O4/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)23-19-10-9-15(22-4)11-17(19)21;5-3(6)1-2-4(7)8/h5-11,14H,12-13H2,1-4H3;1-2H,(H,5,6)(H,7,8)/b;2-1-
Formel vikt 444.1719