Läs mer
rac Methotrimeprazine Sulfoxide, TRC
Produktkod. 30466644
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30466644 1 mg
1 options
Produktkod. 30466644 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2607901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methotrimeprazine Sulfoxide
CAS 1188265-45-3
Molekylformel C19H24N2O2S
Kvantitet 1 mg
Synonym 10H-Phenothiazine-10-propanamine, 2-methoxy-N,N,β-trimethyl-, 5-oxide, 3-(2-Methoxy-5-oxophenothiazin-10-yl)-2-methyl-N,N-dimethylpropan-1-amine, 2-Methoxy-N,N,β-trimethyl-10H-phenothiazine-10-propanamine 5-oxide, 10-[3-(Dimethylamino)-2-methylpropyl]-2-methoxyphenothiazine 5-oxide, rac-Methotrimeprazine sulfoxide, (±)-Levomepromazine sulfoxide
InChI formel InChI=1S/C19H24N2O2S/c1-14(12-20(2)3)13-21-16-7-5-6-8-18(16)24(22)19-10-9-15(23-4)11-17(19)21/h5-11,14H,12-13H2,1-4H3
LEDER COc1ccc2c(c1)N(CC(C)CN(C)C)c3ccccc3S2=O
IUPAC-namn 3-(2-methoxy-5-oxophenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine
Molekylvikt (g/mol) 344.47
Formel vikt 344.1558