Läs mer
rac Methyl Efavirenz (Mixture of Diastereomers), TRC
Produktkod. 30475653
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30475653 1 mg
1 options
Produktkod. 30475653 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3039001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac methyl Efavirenz (mixture of diastereomers)
CAS 353270-76-5
Molekylformel C15 H11 Cl F3 N O2
Kvantitet 1 mg
Synonym 2H-3,1-Benzoxazin-2-one','6-chloro-1,4-dihydro-4-[2-(2-methylcyclopropyl)ethynyl]-4-(trifluoromethyl)-','(4S)-, 2H-3,1-Benzoxazin-2-one','6-chloro-1,4-dihydro-4-[(2-methylcyclopropyl)ethynyl]-4-(trifluoromethyl)-','(4S)- (9CI), (4S)-6-Chloro-1,4-dihydro-4-[2-(2-methylcyclopropyl)ethynyl]-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, Methyl-DMP 266, SV 997
InChI formel InChI=1S/C15H11ClF3NO2/c1-8-6-9(8)4-5-14(15(17,18)19)11-7-10(16)2-3-12(11)20-13(21)22-14/h2-3,7-9H,6H2,1H3,(H,20,21)/t8?,9?,14-/m0/s1
LEDER CC1CC1C#C[C@@]2(OC(=O)Nc3ccc(Cl)cc23)C(F)(F)F
IUPAC-namn (4S)-6-chloro-4-[2-(2-methylcyclopropyl)ethynyl]-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one
Molekylvikt (g/mol) 329.7
Formel vikt 329.043