Läs mer
rac Mono(2-ethyl-5-oxohexyl) Phthalate, TRC
Produktkod. 30040690
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040690 10 mg
30040476 1 mg
30040425 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040690 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM54252010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40321-98-0
Molekylformel C16 H20 O5
Molekylvikt (g/mol) 292.33
Synonym rac Mono(2-ethyl-5-oxohexyl) Phthalate, Phthalic acid mono-2-ethyl-5-oxohexyl ester
IUPAC-namn 2-(2-ethyl-5-oxohexoxy)carbonylbenzoic acid
LEDER CCC(CCC(=O)C)COC(=O)c1ccccc1C(=O)O
Specifikationer
Kemiskt namn eller material Phthalic acid, mono-2-ethyl-5-oxohexyl ester
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C16H20O5/c1-3-12(9-8-11(2)17)10-21-16(20)14-7-5-4-6-13(14)15(18)19/h4-7,12H,3,8-10H2,1-2H3,(H,18,19)
Formel vikt 292.1311
Procent renhet >95%