Läs mer
rac N-Acetyl-Pseudoephedrine, TRC
Produktkod. 30323358
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30323358 250 mg
1 options
Produktkod. 30323358 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA187610250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material rac N-Acetyl-Pseudoephedrine
CAS 5878-95-5
Molekylformel C12H17NO2
Kvantitet 250 mg
Synonym rel-N-[(1S,2S)-2-Hydroxy-1-methyl-2-phenylethyl]-N-methylacetamide, (S*,S*)-(±)-N-(2-Hydroxy-1-methyl-2-phenylethyl)-N-methylacetamide, threo-(±)-N-(β-Hydroxy-α-methylphenethyl)-N-methylacetamide
InChI formel InChI=1S/C12H17NO2/c1-9(13(3)10(2)14)12(15)11-7-5-4-6-8-11/h4-9,12,15H,1-3H3
LEDER OC([C@@H](N(C)C(C)=O)C)C1=CC=CC=C1
IUPAC-namn N-(1-hydroxy-1-phenylpropan-2-yl)-N-methylacetamide
Molekylvikt (g/mol) 207.27
Formel vikt 207.1259